6-Methyl-1,7-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one 97%

Supplier: Ambeed
Warning

A195338-100MG A195338-250MG A195338-5G
76814-054EA 87 USD
76814-054 76814-056 76814-058
6-Methyl-1,7-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one 97%
6-Methyl-1,7-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one

Precaución: for Research Use Only

Formula: C₆H₆N₄O
MW: 150.14 g/mol
MDL Number: MFCD13195854
CAS Number: 30129-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR