2'-Methoxyacetophenone 95%

Supplier: Ambeed
Warning

A194795-100G A194795-10G A194795-1G A194795-25G A194795-5G
76814-028EA 114 USD
76814-028 76814-030 76814-032 76814-034 76814-036
2'-Methoxyacetophenone 95%
2'-Methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD00008725
CAS Number: 579-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR