6-Nitro-m-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-nitrophenol, 1-Hydroxy-3-methyl-6-nitrobenzene

A194739-100G A194739-10G A194739-25G A194739-5G
76814-018EA 51 USD
76814-018 76814-020 76814-022 76814-024
6-Nitro-m-cresol 98%
6-Nitro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MDL Number: MFCD00007111
CAS Number: 700-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR