2,6-Dichloro-5-fluoronicotinamide 98%

Supplier: Ambeed
Warning

A193598-100G A193598-1G A193598-250MG A193598-25G A193598-5G
76813-990EA 85.94 USD
76813-990 76813-992 76813-994 76813-996 76813-998
2,6-Dichloro-5-fluoronicotinamide 98%
2,6-Dichloro-5-fluoronicotinamide

Precaución: for Research Use Only

Formula: C₆H₃Cl₂FN₂O
MDL Number: MFCD01571365
CAS Number: 113237-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR