Methyl 3-amino-5-chloropyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A192516-100MG A192516-250MG A192516-1G
76813-982EA 185 USD
76813-982 76813-984 77671-706
Methyl 3-amino-5-chloropyrazine-2-carboxylate 97%
Methyl 3-amino-5-chloropyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O₂
MDL Number: MFCD11043614
CAS Number: 28643-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR