2-Chlorooxazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A192451-100MG A192451-10G A192451-1G A192451-250MG A192451-5G
76813-972EA 27 USD
76813-972 76813-974 76813-976 76813-978 76813-980
2-Chlorooxazole-4-carboxylic acid 97%
2-Chlorooxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂ClNO₃
MDL Number: MFCD11040385
CAS Number: 706789-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR