(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (3S,4R)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

A192392-100MG A192392-10G A192392-1G A192392-250MG A192392-5G
76813-962EA 44.56 USD
76813-962 76813-964 76813-966 76813-968 76813-970
(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate 97%
(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈FNO₃
MW: 219.26 g/mol
MDL Number: MFCD18632748
CAS Number: 1174020-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR