2-(2-Bromophenyl)propan-2-ol 98%

Supplier: Ambeed
Warning

A192202-1G A192202-250MG A192202-5G
76813-942EA 45.62 USD
76813-942 76813-944 77593-230
2-(2-Bromophenyl)propan-2-ol 98%
2-(2-Bromophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
MDL Number: MFCD00767180
CAS Number: 7073-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR