4-(6-Aminopyridin-3-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A192066-1G A192066-250MG
76813-938EA 378.75 USD
76813-938 76813-940
4-(6-Aminopyridin-3-yl)benzoic acid 95%
4-(6-Aminopyridin-3-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
MDL Number: MFCD06801984
CAS Number: 222986-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR