tert-Butyl ((4-methylpyridin-2-yl)methyl)carbamate 95+%

Supplier: Ambeed
Warning

A192062-100MG A192062-1G A192062-250MG A192062-5G
76813-930EA 15 USD
76813-930 76813-932 76813-934 76813-936
tert-Butyl ((4-methylpyridin-2-yl)methyl)carbamate 95+%
tert-Butyl ((4-methylpyridin-2-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD19981673
CAS Number: 1330755-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR