(4-Butoxy-2-fluorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-N-Butoxy-2-fluorophenylboronic acid

A191665-100MG A191665-1G
76813-920EA 31 USD
76813-920 76813-922
(4-Butoxy-2-fluorophenyl)boronic acid 95%
(4-Butoxy-2-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
MW: 212.03 g/mol
MDL Number: MFCD08701767
CAS Number: 1107603-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR