4-Chloro-6-hydroxynicotinic acid 98%

Supplier: Ambeed
Warning

A191325-1G A191325-250MG A191325-5G
76813-892EA 88 USD
76813-892 76813-894 76813-896
4-Chloro-6-hydroxynicotinic acid 98%
4-Chloro-6-hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MDL Number: MFCD13189223
CAS Number: 73038-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR