Methyl-4-amino-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-1H-pyrazole-3-carboxylate

A190571-100MG A190571-10G A190571-1G A190571-250MG A190571-5G
76813-882EA 32.89 USD
76813-882 76813-884 76813-886 76813-888 76813-890
Methyl-4-amino-1H-pyrazole-3-carboxylate 95%
Methyl-4-amino-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
MDL Number: MFCD02646737
CAS Number: 360056-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR