3-(Hydroxymethyl)-5-methylphenylboronic Acid 98%

Supplier: Ambeed
Warning

A190047-100MG A190047-1G A190047-250MG
76813-876EA 35 USD
76813-876 76813-878 76813-880
3-(Hydroxymethyl)-5-methylphenylboronic Acid 98%
3-(Hydroxymethyl)-5-methylphenylboronic Acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD16295237
CAS Number: 1451391-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR