5-Bromopyrimidine-2-carboxamide 98%

Supplier: Ambeed
Warning

A189568-100MG A189568-1G A189568-250MG
76813-870EA 18.04 USD
76813-870 76813-872 76813-874
5-Bromopyrimidine-2-carboxamide 98%
5-Bromopyrimidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O
MDL Number: MFCD00507192
CAS Number: 38275-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR