4-Bromo-4'-(diphenylamino)biphenyl 97%

Supplier: Ambeed
Warning

A188480-10G A188480-1G A188480-250MG
76813-828EA 89 USD
76813-828 76813-830 77883-402
4-Bromo-4'-(diphenylamino)biphenyl 97%
4-Bromo-4'-(diphenylamino)biphenyl

Precaución: for Research Use Only

Formula: C₂₄H₁₈BrN
MDL Number: MFCD09038499
CAS Number: 202831-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR