6-Chloro-5-(trifluoromethyl)pyridin-2-amine 98%

Supplier: Ambeed
Warning

A188448-100MG A188448-1G A188448-25G A188448-5G A188448-250MG A188448-10G
76813-822EA 13 USD
76813-822 76813-824 76813-826 77567-754 77671-670 77883-401
6-Chloro-5-(trifluoromethyl)pyridin-2-amine 98%
6-Chloro-5-(trifluoromethyl)pyridin-2-amine

Precaución: For research use only.

Formula: C₆H₄ClF₃N₂
MDL Number: MFCD11848573
CAS Number: 79456-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR