2,4-Dichloro-7-nitroquinazoline 95%

Supplier: Ambeed
Warning

A188149-100MG A188149-1G
76813-820EA 50.93 USD
76813-820 77567-752
2,4-Dichloro-7-nitroquinazoline 95%
2,4-Dichloro-7-nitroquinazoline

Precaución: For research use only.

Formula: C₈H₃Cl₂N₃O₂
MW: 244.03 g/mol
MDL Number: MFCD18157742
CAS Number: 129112-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR