3-Fluoro-5-methoxyphenylacetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Cyanomethyl)-5-fluoroanisole, 2-(3-Fluoro-5-methoxyphenyl)acetonitrile, 3-Fluoro-5-methoxybenzyl cyanide

A187984-100MG A187984-1G A187984-250MG
76813-812EA 8.49 USD
76813-812 76813-814 76813-816
3-Fluoro-5-methoxyphenylacetonitrile 98%
3-Fluoro-5-methoxyphenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₈FNO
MW: 165.164 g/mol
MDL Number: MFCD08741379
CAS Number: 914637-31-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR