2-Piperidone 95%

Supplier: Ambeed
Warning

A187204-100G A187204-10G A187204-1G A187204-25G A187204-500G A187204-5G
76813-782EA 43 USD
76813-782 76813-784 76813-786 76813-788 76813-790 76813-792
2-Piperidone 95%
2-Piperidone

Precaución: for Research Use Only

Formula: C₅H₉NO
MDL Number: MFCD00006037
CAS Number: 675-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR