(1S)-1-(2-Fluorophenyl)ethan-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Fluoro-α-methylbenzyl alcohol, (S)-1-(2-Fluorophenyl)ethanol

A186625-1G A186625-250MG A186625-5G A186625-100MG
76813-762EA 37 USD
76813-762 76813-764 76813-766 77883-392
(1S)-1-(2-Fluorophenyl)ethan-1-ol 97%
(1S)-1-(2-Fluorophenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉FO
MDL Number: MFCD03092992
CAS Number: 171032-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR