3-Bromo-5-fluorobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A186589-100MG A186589-1G A186589-250MG A186589-5G
76813-754EA 21.22 USD
76813-754 76813-756 76813-758 76813-760
3-Bromo-5-fluorobenzene-1-sulfonyl chloride 95%
3-Bromo-5-fluorobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃BrClFO₂S
MW: 273.507 g/mol
MDL Number: MFCD13185364
CAS Number: 1214342-44-5
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR