H-β-HoPhe-OH 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Amino-4-phenylbutyric acid, L-β-Homophenylalanine

A186567-5G A186567-1G
76813-752EA 297.06 USD
76813-752 77671-636
H-β-HoPhe-OH 97%
H-β-HoPhe-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
MDL Number: MFCD03428441
CAS Number: 26250-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR