N,N-Dimethyl-4-nitrobenzylamine 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-1-(4-nitrophenyl)methanamine

A185942-10G A185942-1G A185942-25G
76813-742EA 124.13 USD
76813-742 76813-744 76813-746
N,N-Dimethyl-4-nitrobenzylamine 95%
N,N-Dimethyl-4-nitrobenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.204 g/mol
MDL Number: MFCD00035956
CAS Number: 15184-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR