1-(1-Methylpiperidin-4-yl)-1H-pyrazol-4-amine 97%

Supplier: Ambeed
Warning

A185614-1G A185614-5G A185614-250MG
76813-726EA 248 USD
76813-726 76813-728 77567-706
1-(1-Methylpiperidin-4-yl)-1H-pyrazol-4-amine 97%
1-(1-Methylpiperidin-4-yl)-1H-pyrazol-4-amine

Precaución: For research use only.

Formula: C₉H₁₆N₄
MDL Number: MFCD19704303
CAS Number: 1201935-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR