7-Fluoro-2,3-dihydroinden-1-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Fluoroindan-1-one
A184912-100MG
A184912-10G
A184912-1G
A184912-250MG
A184912-25G
A184912-5G
76813-702EA
12.74
USD
76813-702
76813-704
76813-706
76813-708
76813-710
76813-712
7-Fluoro-2,3-dihydroinden-1-one 98%
7-Fluoro-2,3-dihydroinden-1-one
Precaución: for Research Use Only
Formula:
C₉H₇FO MW: 150.1500 g/mol |
MDL Number:
MFCD07368749 CAS Number: 651735-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...