exo-8-Azabicyclo[3.2.1]octan-3-ol hydrochloride 97%

Supplier: Ambeed
Warning

A184472-250MG A184472-1G
76813-698EA 35 USD
76813-698 77671-610
exo-8-Azabicyclo[3.2.1]octan-3-ol hydrochloride 97%
exo-8-Azabicyclo[3.2.1]octan-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO
MDL Number: MFCD21642368
CAS Number: 17366-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR