Methyl-1-methyl-1H-pyrazole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-pyrazole-4-carboxylate

A184464-1G A184464-250MG A184464-25G A184464-5G
76813-690EA 16.98 USD
76813-690 76813-692 76813-694 76813-696
Methyl-1-methyl-1H-pyrazole-4-carboxylate 98%
Methyl-1-methyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD11053755
CAS Number: 5952-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR