Ethyl 3-chlorothieno[2,3-b]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A184303-1G A184303-250MG
76813-686EA 98.67 USD
76813-686 76813-688
Ethyl 3-chlorothieno[2,3-b]pyridine-2-carboxylate 95%
Ethyl 3-chlorothieno[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂S
MW: 241.69 g/mol
MDL Number: MFCD08695791
CAS Number: 66869-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR