2,5-Dibromofuran 99% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabiised with magnesium oxide (MgO)

A183367-1G A183367-5G
76813-642EA 57 USD
76813-642 76813-644
2,5-Dibromofuran 99% stabilized
2,5-Dibromofuran

Precaución: for Research Use Only

Formula: C₄H₂Br₂O
MDL Number: MFCD00191342
CAS Number: 32460-00-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR