2-Methoxy-5-nitrophenol 98%

Supplier: Ambeed
Warning

A182065-100G A182065-25G A182065-5G A182065-500G
76813-620EA 26 USD
76813-620 76813-622 76813-624 76813-626 77671-572
2-Methoxy-5-nitrophenol 98%
2-Methoxy-5-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₇NO₄
MDL Number: MFCD00015561
CAS Number: 636-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR