5,5-Dimethyloxazolidine-2,4-dione (dimethadione) 95%

Supplier: Ambeed
Warning

A181810-100G A181810-10G A181810-1G A181810-25G A181810-5G A181810-500G
76813-588EA 43 USD
76813-588 76813-590 76813-592 76813-594 76813-596 77671-568
5,5-Dimethyloxazolidine-2,4-dione (dimethadione) 95%
5,5-Dimethyloxazolidine-2,4-dione (dimethadione)

Precaución: for Research Use Only

Formula: C₅H₇NO₃
MDL Number: MFCD00005379
CAS Number: 695-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR