2-Iodo-1,4-benzenediamine 95%

Supplier: Ambeed
Warning

A181201-1G A181201-250MG A181201-5G
76813-568EA 22.28 USD
76813-568 76813-570 76813-572
2-Iodo-1,4-benzenediamine 95%
2-Iodo-1,4-benzenediamine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MW: 234.04 g/mol
MDL Number: MFCD04971329
CAS Number: 69951-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR