4-Cyano-6-azaindole 95%

Supplier: Ambeed
Danger

Synonyms: 1H-Pyrrolo[2,3-c]pyridine-4-carbonitrile

A181014-1G A181014-250MG
76813-564EA 82 USD
76813-564 76813-566
4-Cyano-6-azaindole 95%
4-Cyano-6-azaindole

Precaución: for Research Use Only

Formula: C₈H₅N₃
MDL Number: MFCD12963363
CAS Number: 1190319-59-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR