3-Benzyl-6-bromo-2-methoxyquinoline 95%

Supplier: Ambeed
Warning

Synonyms: Bedaquiline Impurity 5

A180757-100G A180757-1G A180757-250MG A180757-25G A180757-5G
76813-554EA 277 USD
76813-554 76813-556 76813-558 76813-560 76813-562
3-Benzyl-6-bromo-2-methoxyquinoline 95%
3-Benzyl-6-bromo-2-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₇H₁₄BrNO
MW: 328.20 g/mol
MDL Number: MFCD22493488
CAS Number: 654655-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR