1-Phenyl-1H-pyrazole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A180750-100MG A180750-1G A180750-250MG
76813-548EA 91.24 USD
76813-548 76813-550 76813-552
1-Phenyl-1H-pyrazole-3-carbaldehyde 95%
1-Phenyl-1H-pyrazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
MDL Number: MFCD08437287
CAS Number: 40261-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR