Ethyl 3-chloro-4-formylbenzoate 98%

Supplier: Ambeed
Warning

A180380-1G A180380-250MG A180380-5G
76813-534EA 115.64 USD
76813-534 76813-536 76813-538
Ethyl 3-chloro-4-formylbenzoate 98%
Ethyl 3-chloro-4-formylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
MW: 212.63 g/mol
MDL Number: MFCD21603681
CAS Number: 1465327-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR