2-Chloro-4,6-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A179973-100MG A179973-250MG A179973-5G A179973-1G
76813-522EA 6 USD
76813-522 76813-524 77611-742 77883-370
2-Chloro-4,6-difluorobenzoic acid 97%
2-Chloro-4,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MDL Number: MFCD16250104
CAS Number: 1242339-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR