tert-Butyl 5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-hydroxy-2-aza-bicyclo[2.2.1]heptane-2-carboxylate

A179947-100MG A179947-1G A179947-5G
76813-520EA 22.28 USD
76813-520 77593-142 77671-544
tert-Butyl 5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate 97%
tert-Butyl 5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.273 g/mol
MDL Number: MFCD11865204
CAS Number: 207405-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR