5-Chloroisoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophthalimide

A179304-100MG A179304-10G A179304-1G A179304-5G A179304-250MG
76813-504EA 13.8 USD
76813-504 76813-506 76813-508 76813-510 77671-540
5-Chloroisoindoline-1,3-dione 98%
5-Chloroisoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C8H4ClNO2
MW: 181.5759 g/mol
MDL Number: MFCD08703319
CAS Number: 7147-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR