2-Bromo-4-chlorophenol 98%
Supplier: Ambeed
|
Warning
|
A178962-1000G
A178962-100G
A178962-25G
A178962-500G
A178962-5G
76813-494EA
352
USD
76813-494
76813-496
76813-498
76813-500
76813-502
2-Bromo-4-chlorophenol 98%
2-Bromo-4-chlorophenol
Precaución: for Research Use Only
Formula:
C₆H₄BrClO |
MDL Number:
MFCD00002319 CAS Number: 695-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...