(R)-Chroman-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

A178396-100MG A178396-250MG A178396-1G A178396-5G
76813-482EA 11 USD
76813-482 76813-484 77671-526 77881-351
(R)-Chroman-4-amine hydrochloride 95%
(R)-Chroman-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MDL Number: MFCD08457629
CAS Number: 730980-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR