2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-diethylamino-2'-hydroxycarbonylbenzophenone, 2-(4-(Diethylamino)-2-hydroxybenzoyl)benzoic acid

A178376-1000G A178376-100G A178376-1G A178376-25G A178376-500G A178376-5G
76813-470EA 154.9 USD
76813-470 76813-472 76813-474 76813-476 76813-478 76813-480
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid 98%
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₄
MW: 313.35 g/mol
MDL Number: MFCD00134669
CAS Number: 5809-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR