(R)-3-(((Benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Dap(Cbz)-OH

A178351-1G A178351-25G A178351-5G
76813-464EA 45 USD
76813-464 76813-466 76813-468
(R)-3-(((Benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
(R)-3-(((Benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₆
MDL Number: MFCD18252750
CAS Number: 81306-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR