3-Methyl-1H-pyrazole-4-carbonitrile 98%

Supplier: Ambeed
Warning

A178308-100MG A178308-1G A178308-250MG A178308-5G
76813-456EA 27.59 USD
76813-456 76813-458 76813-460 76813-462
3-Methyl-1H-pyrazole-4-carbonitrile 98%
3-Methyl-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₅N₃
MW: 107.11 g/mol
MDL Number: MFCD05664572
CAS Number: 131661-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR