EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
EMCH, 3,3'-N-[ε-Maleimidocaproic acid] hydrazide hydrochloride, EMCH.HCl, 6-Maleimidohexanehydrazide hydrochloride, E-Maleimidocaproic acid hydrazine hydrochloride, N-ε-Maleimidocaproic acid hydrazide hydrochloride
A177662-100MG
A177662-1G
A177662-250MG
A177662-5G
76813-426EA
14
USD
76813-426
76813-428
76813-430
77883-350
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride) 95%
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N₃O₃·ClH |
MDL Number:
MFCD07363434 CAS Number: 175290-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...