1,3-Dibromo-5-isopropoxybenzene 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromoisopropoxybenzene

A177265-1G A177265-250MG A177265-5G
76813-420EA 6 USD
76813-420 76813-422 76813-424
1,3-Dibromo-5-isopropoxybenzene 97%
1,3-Dibromo-5-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂O
MDL Number: MFCD12547881
CAS Number: 1112210-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR