Ethyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A177198-250MG A177198-1G
76813-418EA 78.51 USD
76813-418 77671-516
Ethyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate 97%
Ethyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD07388488
CAS Number: 70500-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR