(3-Bromo-5-isopropoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-isopropoxyphenylboronic acid

A177165-1G A177165-250MG
76813-414EA 14.86 USD
76813-414 76813-416
(3-Bromo-5-isopropoxyphenyl)boronic acid 97%
(3-Bromo-5-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BBrO₃
MW: 258.91 g/mol
MDL Number: MFCD07784398
CAS Number: 871125-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR