2-Aminothiazol-4-ol 95%

Supplier: Ambeed
Warning

A177015-10G A177015-1G A177015-5G A177015-100G
76813-408EA 39 USD
76813-408 76813-410 76813-412 77671-512
2-Aminothiazol-4-ol 95%
2-Aminothiazol-4-ol

Precaución: for Research Use Only

Formula: C₃H₄N₂OS
MDL Number: MFCD08692660
CAS Number: 7146-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR